LinerGOLD™ GC Liners
Supplier: Thermo Scientific
453A2265-UI 453A1262-UI 453A1295-UI 453A2292-UI 453A1345-UI 453A2342-UI 453A2922-UI 453A1355-UI 453A2352-UI 453A1255-UI 453A1252-UI 453A1335-UI 45350032-UI 45350033-UI 453T2845-UI 45350030-UI 453T1905-UI 453T2999-UI 453A0415 453A0411
10055-314EA 385.21 CAD
10055-314 10055-316 10055-318 10055-320 10055-322 10055-324 10056-260 10056-262 10056-264 10056-270 10056-274 10056-276 10056-278 10056-280 10056-286 10056-288 10056-290 10056-292 10056-308 10056-310 10056-312 10056-314 10056-316 10056-878 10056-882 10056-886 10056-888 10056-890 10171-634 10171-636 10171-640 10171-642 10171-646 10171-648 10171-654 10171-658 10171-660 10171-670 10171-672 10171-676 10171-688 10171-694 10171-700 10171-718 10171-724 10171-730 76207-554 76207-556
LinerGOLD™ GC Liners
Chromatography Supplies Chromatography Inlet Liners
Use the gold standard in GC liner performance and consistency. Thermo Scientific™ LinerGOLD™ GC Liners provide enhanced inertness, leading to increased accuracy, sensitivity, and precision in your gas chromatography (GC) analysis.
  • Increased accuracy and sensitivity in analysis, preventing the loss of sensitive compounds in analysis
  • Lower detection limits when analyzing active and sensitive compounds
  • Excellent reproducibility gives confidence in your results from liner to liner
  • Consistently high level of performance time after time
LinerGOLD GC liners are manufactured and tested to ensure that you have confidence with every liner that is installed onto your GC system.