Thermo Scientific Super Clean Cartridge Filter Baseplates, Thermo Scientific
Supplier: Thermo Scientific
60180-814 60180-815 60180-817 60180-818
10055-288EA 1001.22 CAD
10055-288 10055-290 10055-296 10055-302
Thermo Scientific Super Clean Cartridge Filter Baseplates, Thermo Scientific
Filters In-Line Filters
A range of baseplate options allows configuration to your individual requirements. Needle valves ensure that the gas line is not contaminated during cartridge change.
Baseplates require one-time installation only.
Delivery information: Super Clean Cartridge Filters are not included.