HyperREZ™ XP Carboyhydrate Ca²⁺ HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
69708-254080 69208-057726 69208-307780 69208-903027
10043-070EA 3634.05 CAD
10043-070 10046-252 10046-254 10046-256
HyperREZ™ XP Carboyhydrate Ca²⁺ HPLC Columns, Thermo Scientific
Chromatography Columns
HyperREZ XP Carbohydrate columns are based on a monodisperse resin with a 4 or 8% divinylbenzene content, and provide an ideal medium for the analysis of carbohydrates and organic acids.
Unlike silica based columns they are stable at low pH, allowing the use of dilute acid as a mobile phase. The columns can also be run at elevated temperatures, for faster analysis and improved resolution of some closely eluting analytes.