ProPac™ PA1 Protein and Peptide Columns, Thermo Scientific
Supplier: Thermo Scientific
039658 040137 039657
10040-510EA 6182.4 CAD
10040-510 10040-512 10045-854
ProPac™ PA1 Protein and Peptide Columns, Thermo Scientific
Chromatography Columns
Efficiently analyze and purify hydrophilic anionic proteins and peptides with the Thermo Scientific™ ProPac PA1 Protein and Peptide column.
These strong anion-exchange columns provide high-resolution separations of proteins with pI values from 3 to 11.