Lab Sinks, Self Rimming, Two Bowl, V-1 Series, Eagle MHC
Supplier: Eagle Group
SR10B2V1 SR12A2V1 SR14D2V1 SR16B2V1 SR16C2V1 SR18A2V1 SR22A2V1 SR24B2V1
10033-218EA 3232.29 CAD
10033-218 10033-220 10033-222 10033-336 10033-338 10033-340 10033-342 10033-344
Lab Sinks, Self Rimming, Two Bowl, V-1 Series, Eagle MHC
Furniture Casework
Heavy gauge type 304 series stainless steel coved bowls with large radius.
  • Two compartment
  • Self rimming, deep drawn design
1¹/₂"diameter basket drain in center of bowl. Sink can be mounted on top or bottom of countertop. Features 1³/₈" hole for mixing faucet.
Order Now
SPECIFICATIONS
Environmentally Preferable