Neurotransmitter Transporter Uptake Assay Kit, Molecular Devices
Supplier: MOLECULAR DEVICES LLC. CA
R6138 R8173 R8174 R6138 R8173 R8174
10048-832EA 364.05 CAD
10048-832 10048-834 10048-836 CA10048-832 CA10048-834 CA10048-836
Neurotransmitter Transporter Uptake Assay Kit, Molecular Devices
Assays Cellular Assays
Neurotransmitter transporters play a key role in depression and neurodegenerative diseases (NDD) such as Alzheimer's and Parkinson's.
  • Replaces radioactive assays
  • One-step, homogeneous assay format
  • Two assay setup
The ability to monitor serotonin, norepinephrine and dopamine neurotransmitter uptake is key to a better understanding of these diseases.
Homogeneous, live-cell fluorescent assay replaces radioactive assays, reducing disposal costs and eliminating safety risks associated with radiolabeled assays. One-step, homogeneous assay format is amenable to automation and high-throughput screening. Two assay setup options offer greater flexibility: endpoint mode for high-throughput applications, or real-time kinetic mode for mechanistic studies.