TraceGOLD TG-5MS GC Columns, Thermo Scientific
Supplier: Thermo Scientific
26098-5820 26098-5780 26098-4100 26098-2760 26098-2940
10038-936EA 1348.47 CAD
10038-936 10038-938 10038-940 10038-944 10038-950 10038-952 10038-954 10038-956 10038-958 10038-960 10038-962 10038-964 10038-966 10038-968 10038-970 10038-972 10038-974 10038-976 10038-980 10038-982 10038-984 10038-986 10038-988 10038-990 10038-992 10038-994 10056-720 10056-728
TraceGOLD TG-5MS GC Columns, Thermo Scientific
Chromatography Columns
Employ the most widely used MS phase in gas chromatography with the 5% phenyl phase Thermo Scientific TraceGOLD TG-5MS GC Column.
Low polarity phase, 5% diphenyl/95% dimethyl polysiloxane. Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity. Exceptional inertness ideal for analysis of active compounds. Equivalent to USP G27 phase. Similar to Rxi-5ms, DB-5, HP-5, HP-5ms, Ultra-2, SPB-5, Equity-5, CP-Sil 8. Recommended for semivolatiles, phenols, amines, residual solvents and solvent impurities, drugs of abuse, pesticides, PCB cogeners, and aroclor mixes.