TraceGOLD™ GC Columns with SafeGuard, Thermo Scientific
Supplier: Thermo Scientific
26088-1421 26088-1435 26096-1301 26096-1421 26096-1425 26096-2235 26098-0475 26098-1305 26098-1425 26098-1435 26098-5785 26099-1425 26099-2985
10055-730EA 2029.61 CAD
10055-730 10056-388 10056-426 10056-604 10056-606 10056-616 10056-676 10056-680 10056-688 10056-692 10056-746 10056-758 10056-800
TraceGOLD™ GC Columns with SafeGuard, Thermo Scientific
Chromatography Columns
Extend your column lifetime without compromising performance.
SafeGuard is an integrated guard on the GC column.