Incu-Mixer™ Microplate vortexer, Benchmark Scientific
Supplier: BENCHMARK SCIENTIFIC CA
H6002 H6004
490005-874EA 7292.74 CAD
490005-874 490015-278 CA76323-982 CA76323-984
Incu-Mixer™ Microplate vortexer, Benchmark Scientific
Shakers Vortex Mixers
The new Incu-Mixer™ MP series is designed to provide an optimal method for vortexing and incubating microplates. Simultaneous heating from both the bottom surface and the lid provide temperature uniformity throughout the sample. Additionally, the heated lid minimizes evaporation and prevents condensation.
  • Optimized for incubated mixing in microplates
  • Heated lid prevents condensation
  • Accepts plates up to 40 mm high
  • Independently control time, temperature and speed
  • Available in two-plate or four-plate format
A compact, 2 mm orbit ensures thorough mixingMore Product Information