Secura® Balances, Standard Models, 100 - 240 V, Sartorius
Supplier: Sartorius
SECURA125-1S SECURA225D-1S
CA101485-972EA 5749.64 CAD
CA101485-972 CA101485-974 CA101485-976 CA101485-978 CA101485-980 CA101485-982 CA101485-984 CA101485-986 CA101486-028 CA102355-764 CA102355-796 CA102355-798 CA490019-378 CA490019-380 CA76412-436 CA76412-452 CA76412-458 CA76412-464
Secura® Balances, Standard Models, 100 - 240 V, Sartorius
Balances and Scales
Secura is a laboratory balance that does far more than set new standards in metrological specifications.
  • LevelControl Keeps Balance Correctly Leveled
  • No Risk Due to Temperature Fluctuations
  • Stress-Free USP Compliant Measurements
  • Password Protection
  • Interactive User Guidance for Fast GLP and GMP-Compliant Printouts
It also features built-in protection systems for complete reliability and regulatory compliance.
Real-time guidance prompts for leveling, automatic internal adjustment, and 100% traceable, clear documentation with sample and batch identifiers make lab work more efficient.