BioClave™ Digital Benchtop Autoclaves, Benchmark Scientific
Supplier: BENCHMARK SCIENTIFIC CA
B4000-16 B4000-M
490012-494EA 16853.45 CAD
490012-494 CA76323-980 CA76333-882
BioClave™ Digital Benchtop Autoclaves, Benchmark Scientific
Sterilizers Autoclaves (Steam Sterilizers)
Steam sterilization of research tools and consumables is convenient and affordable with the new BioClave digital research autoclaves. With a width of under 18 (under 14 for the BioClave Mini) both models will fit comfortably on the most crowded bench-top.
  • Mechanical and electrical safety interlock engages until the pressure has reached zero PSI
  • Three trays (2 with the BioClave Mini)
  • Fully automatic operation, all segments of the sterilization cycle (fill, sterilize, exhaust, and dry) commence and run to completion by simply pressing the start button
  • Mini printer is also available for BioClave 16 providing record of sterilization parameters
The 16 L BioClave offers three cycle options (standard programs) which are preset for the sterilization of liquids, wrapped instruments, or unwrapped instruments/plastic-ware. A fourth option dry only is also available for additional drying time to be added to the end of the cycle. For special applications, requiring variations in cycle parameters, all settings are fully adjustable.
The 8 L BioClave Mini is preprogrammed to meet basic sterilization needs. Although the temperature can be selected at either 121 °C or 134 °C, the sterilization time is fixed. The economical BioClave Mini is the ideal choice wherever bench-top sterilization is required and space is limited.
Delivery information: One year warranty.
Caution: Article 490012-494 is 120 V, 60 Hz, if a unit suitable for use with European voltage (240 V, 50 Hz) is required, please order 470230-604