Labels, Diversified Biotech
Supplier: Diversified Biotech
RNBW-1000 RNBW-1100 SPOT-1000 SPOT-1100 SPOT-1200 SPOT-1300 SPOT-1400 SPOT-1500 SPOT-2000 SPOT-2100 SPOT-2200 SPOT-2300 SPOT-2400 SPOT-2500 SPOT-3000 SPOT-5000 T-SPOTS T-SPOTS-100 T-SPOTS-50 T-SPOTS-50B T-SPOTS-50D T-SPOTS-50G T-SPOTS-50L T-SPOTS-50O T-SPOTS-50P T-SPOTS-50R T-SPOTS-50S T-SPOTS-50Y T-SPOTS-75 T-SPOTS-B T-SPOTS-C T-SPOTS-D T-SPOTS-G T-SPOTS-L T-SPOTS-O T-SPOTS-P T-SPOTS-R T-SPOTS-S T-SPOTS-Y T-SPOTS-50C SPOT-4000 TS-TNY T-SPOTS-44 T-SPOTS-M
490006-916EA 233.32 CAD
490006-916 490006-918 490006-940 490006-942 490006-944 490006-946 490006-948 490006-950 490006-952 490006-954 490006-956 490006-958 490006-960 490006-962 490006-964 490006-966 490007-008 490007-010 490007-012 490007-014 490007-016 490007-018 490007-020 490007-022 490007-024 490007-026 490007-028 490007-030 490007-032 490007-034 490007-036 490007-038 490007-040 490007-042 490007-044 490007-046 490007-048 490007-050 490007-052 490014-294 490014-300 10141-736 10141-738 10141-900
Labels, Diversified Biotech
Labels
Tough-Spots will adhere to all plastics.
  • Chemically inert polyvinyl
  • Temperature resistant from –196 to 80 °C or –196 to 100 °C
  • Pre-cut, peel-off, round labels sized to fit 0.5 to 2.0 ml microcentrifuge tubes and other containers
Tough-Spots withstand boiling water baths, conventional and cryogenic freezer temperatures, most organic solvents, caustic agents and other challenges without peeling, becoming brittle or degrading. Labels can be printed directly from the laser printer.