Miscellaneous Label Applications, Brady
Supplier: Avantor
CA80090-406EA 140.08 CAD
CA80090-406 CA80090-410 CA80090-418 CA80090-420 CA80090-436 CA80090-440 CA80090-442
Miscellaneous Label Applications, Brady
Labels
B-488: High Performance Polyester withstands numerous solvents and variable temperatures when applied to various surfaces including Stainless Steel and Polypropylene.
B-498: Permanent or repositionable, once applied, CleanLift® Series Vinyl Cloth is there to stay unless you want to change it. The adhesive and cloth backing offers excellent holding power, yet allow for clean removal and repositioning.
B-427: Self-Laminating Vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing. Durable, permanent acrylic adhesive has outstanding water and oil resistance.
B-351: Tamper-Resistant Vinyl is designed to fracture easily in order to show signs of product tampering and to prevent on-piece label removal.
Chemical Resistant Lab: Polyester
Created specifically for labeling lsides, this material offers permanent adhesion, outstanding chemical resistance, and write-on capabilities.
Vinyl Material:
Pliable and durable, this material is designed to wrap around and adhere to small diameter curved surfaces, such as straws. The clear "tail" wraps over the text for added protection.
Vinyl Cloth Materials:
This label, with removable adhesive, is easy to handle with an aggressive tack that makes it ideal for use on glass flasks, plastic jugs, bottles, goblets, rough surfaces, and general purpose lab labeling.
Certifications: RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved.