Cryo Labels, Brady
Supplier: Avantor
CA80090-414EA 98.87 CAD
CA80090-414 CA80090-416 CA80090-422 CA80090-600 CA80090-610 CA11011-028
Cryo Labels, Brady
Labels
Durable Polyester (B-490) markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
  • White polyester material has a permanent acrylic adhesive and is thermal transfer printable
  • All-in-one cartridge includes label supply and R4300 Series ribbon