Labels, Brady
Supplier: Avantor
CA80090-082EA 91.15 CAD
CA80090-082 CA80090-096 CA80090-098 CA80090-100 CA80090-102 CA80090-124 CA80090-128 CA80090-130 CA80090-134 CA80090-138 CA80090-142 CA80090-144 CA80090-146 CA80090-150 CA80090-166 CA80090-172 CA80090-196
Labels, Brady
Labels
B-499: Nylon Cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable. This material has a permanent acrylic adhesive and is thermal transfer printable.
B-488: High Performance Polyester withstands numerous solvents and variable temperatures when applied to various surfaces including Stainless Steel and Polypropylene.
B-461: Self-Laminating Polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-427: Self-Laminating Vinyl is an excellent material forMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable