Cryo / Liquid Nitrogen Labels, Laser Printable, Brady
Supplier: Brady Worldwide
LAT-58-361-1 LAT-59-361-2.5 LAT-60-799-2.5
CA80090-324EA 414.65 CAD
CA80090-324 490006-132 490006-134
Cryo / Liquid Nitrogen Labels, Laser Printable, Brady
Labels
Use your laser printer to create labels that withstand extreme laboratory environments including liquid nitrogen and freezers.
Specially designed for laboratory applications, Brady labels guarantee maximum quality, endurance, and resistance against UV exposure, chemicals, solvents, abrasion, and extreme temperatures.
With a full range of standard and custom label sizes, materials, and adhesives, we can provide the exact label to meet your needs. Chemical, solvent, and detergent-resistant labels that can withstand exposure to liquid nitrogen, freezer storage, and the autoclave process.