Dynalon Polypropylene Beakers, Griffin Style
Supplier: Dynalon
222075-5000 222075-0250 222075-0100 222075-0500 222075-0050
13914-507CS 529.54 CAD
13914-507 30620-128 30620-994 30620-996 490007-194
Dynalon Polypropylene Beakers, Griffin Style
Beakers
High-quality beakers are made from polypropylene for all routine laboratory requirements. Graduations molded in milliliters.
  • Beakers are autoclavable to 121°C
  • Special no-drip pouring spout
  • Shatter resistant
Chemically resistant to most acids, bases and many common solvents.