Duran Media Storage Glass Bottles, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
218012458 218013651 218014459
490009-880CS 269.06 CAD
490009-880 490013-166 490013-168
Duran Media Storage Glass Bottles, WHEATON®, DWK Life Sciences
Bottles Cell Culture Bottles
Screw-cap glass media bottles are stable and chemically resistant.
  • Chemically resistant
  • Plastic pouring ring
  • Interchangeable caps and rings
They are complete with a plastic pouring ring which imparts drop-free working. Plastic caps and pouring rings are completely interchangeable since all bottles from 100 ml on have the same thread size. Bottle, pouring ring and cap can be sterilized (up to 140 ˚C).