Supermax Vibrant™ Latex Gloves
Supplier: SUPERMAX HEALTHCARE CANADA CA
98225 98226 98227 98228
CA10500-610CS 291.2 CAD
CA10500-610 CA10500-612 CA10500-614 CA10500-632
Supermax Vibrant™ Latex Gloves
Gloves
Supermax Vibrant™ micro-textured powder-free latex examination gloves are made from 100% natural rubber.
  • Ambidextrous, 5.0 mil palm thickness
  • Micro-textured finish delivers improved grip
  • Advanced rinsing process improves quality and purity
  • Powder-free, non sterile, single-use gloves
Each batch of gloves is air-tested to detect pinholes. Aurelia gloves conform and are tested to the highest international standards.
Packaging: 100 gloves per box.