Drummond Digital Positive Displacement Microdispenser
Supplier: Drummond
3-000-510 3-000-525 3-000-550 3-000-575 3-000-590 3-000-210-G 3-000-225-G 3-000-250-G 3-000-275-G
53507-006PK 1702.51 CAD
53507-006 53507-008 53507-010 53507-012 53507-014 490012-674 490012-676 490012-678 490012-680
Drummond Digital Positive Displacement Microdispenser
Pipettes Micropipettes
Available in five models, this pipettor features a direct digital set and readout for fractional microliters.
  • Positive lock prevents accidental volume changes
  • Bores fit on 1000 µl unit
  • Direct digital set
  • Readout for fractional microliters
  • Five models available
A positive lock prevents accidental volume changes.
The autoclavable pipettor features an overall volumetric accuracy of ±1%.
Capillary glass bores and polypropylene bores fit on 1000 µl unit.
Ordering information: One pack includes 100 microdispensers.