Kimble Chase Glass Pasteur Pipettes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
63B92 63B92P 63A53 63A53P 63C49
89399-924CS 279.22 CAD
89399-924 89399-926 89511-294 89511-298 490012-626 77321-370
Kimble Chase Glass Pasteur Pipettes, DWK Life Sciences
Pipettes Pasteur/Transfer Pipettes
Pasteur Pipettes - Borosilicate and Flint Glass: Manufactured from the highest quality glass, these disposable pipettes feature uniform wall weight and tip diameters.
Packaged in easel dispenser box. Pasteur Pipettes - Cotton plugged: Manufactured from the highest quality glass, cotton plugged pipettes feature uniform wall weight and tip diameters. Extra deep constrictions keep cotton plug from moving. A great time saver for every laboratory. Pasteur Pipettes - Controlled Drop: Controlled drop pipette will accurately and consistently dispense 25 ±3 drops per ml of serum or red cells at 23 °C when held at a 45° angle.