Diversified Biotech Reagent Reservoirs
Supplier: Diversified Biotech
RESE-2000 RESE-3100 RESE-3050 RESE-5000 RESE-5050 RESE-1000 RESE-1050 RESE-2050 RESE-3000
490006-896CS 329.75 CAD
490006-896 490006-900 10141-824 10141-826 10141-828 10141-916 10141-918 10141-920 10141-922
Diversified Biotech Reagent Reservoirs
Reagent Reservoirs
Disposable reservoirs available in sterile and nonsterile formats.
  • Unique trough within a trough maximizes reagent recovery
  • Convenient pour-off spouts on all four corners
  • Made from high impact polystyrene
  • A wide base provides stability and avoids spills
  • Graduated marks on inside wall
  • Superior design provides stronger sidewalls
  • Sterile versions are sterilized by gamma irradiation
The 25mL divider allows for pipetting with eight channels on one side and four channels or a single channel on the other; the divider minimizes reagent loss and is convenient for adding controls.