Eppendorf® Research® Plus Mechanical Single-Channel Pipettes - Trade-In
Supplier: Eppendorf
2231300033 2231300034 2231300035 2231300036 2231300037 2231300038 2231300039 2231300040 2231300041 2231300042
CA490013-018EA 606.84 CAD
CA490013-018 CA490013-020 CA490013-022 CA490013-024 CA490013-026 CA490013-028 CA490013-030 CA490013-032 CA490013-034 CA490013-036
Eppendorf® Research® Plus Mechanical Single-Channel Pipettes - Trade-In
Pipettes Micropipettes
Take advantage and trade in ANY brand pipette for a NEW Eppendorf® Research® plus single-channel variable-volume Pipette at a reduced price. Trust Eppendorf's premium products for quality in your laboratory, backed by 75 years of liquid handling experience and green, cutting-edge manufacturing.
  • Volume adjustment feature allows desired volume to be reached in few quick turns
  • Control button and ejectors low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume display
  • Calibration display and adjustment opening for specific liquids
More Product Information