Neogard® Powder-Free Chloroprene Examination Gloves, Microflex®
Supplier: Microflex
C520 C524 C522 C523 C521
CA10837-056CS 434.55 CAD
CA10837-056 CA10118-980 CA10159-878 CA10159-880 CA10160-604
Neogard® Powder-Free Chloroprene Examination Gloves, Microflex®
Gloves
The Neogard glove is made from a unique, non latex formulation that delivers outstanding grip in wet and dry environments.
  • Ambidextrous
  • Beaded cuffs
  • Textured fingers
With unparalleled tactile sensitivity and strength, Neogard gloves are ideal for every task. Textured fingers ensure a strong and secure grip, while the advanced formulation helps deliver excellent wet and dry grip.