Sartorius Vivaspin® Turbo 15
Supplier: Sartorius
VS15T01 VS15T02 VS15T11 VS15T12 VS15T21 VS15T22 VS15T31 VS15T32 VS15T41 VS15T42 VS15T91 VS15T92
490009-876PK 388.54 CAD
490009-876 490013-972 490013-974 490013-978 490013-980 490013-982 490013-984 490013-986 CA76409-042 CA76409-044 CA76409-046 CA76409-048 CA76409-050 CA76409-052 CA76409-054 CA76409-056 CA76409-058 CA76409-060 CA76409-062 CA76409-064
Sartorius Vivaspin® Turbo 15
Filters Centrifugal Filters
The Vivaspin® Turbo 15 can handle up to 15 ml sample volume in swing bucket rotors and 11 ml in fixed angle rotors accepting 50 ml centrifuge tubes.
  • High chemical resistance
  • High retentate recovery: more than 95%
  • Dead stop volume: 60
  • High flux PES membrane
  • Universal rotor compatibility
The UV joining technology creates a smooth joint transition between membrane and plastic housing, which allows the collection of the complete concentrated sample into the pipette-friendly dead stop pocket.
The unit features a polystyrene butadiene copolymer body and a polyethersulfone membrane with a polystyrene cap.
The Vivaspin® Turbo 15’s sleek internal profile ensures maximum process speeds down to the last few microliters after 100-fold concentration.