picoLUCENT™ PLUS ECL Reagent for Western blotting, G-Biosciences
Supplier: G-Biosciences
786-09AP 786-002 786-09 786-09T 786-09APT 786-165 786-264 786-424 786-165 786-264 786-424
CA95043-394EA 1093.53 CAD
CA95043-394 CA89167-700 CA95043-378 CA95043-380 CA95043-396 10063-116 10063-118 10063-124 CA10118-298 CA10118-300 CA10118-306
picoLUCENT™ PLUS ECL Reagent for Western blotting, G-Biosciences
Assays ELISAs
picoLUCENT™ PLUS-HRP kit is based on our ultra sensitive Luminol substrate that produces chemiluminescence upon reaction with horseradish peroxidase
picoLUCENT™ PLUS-HRP reagents are available in five sizes suitable for 5, 25, 50, 125 and 250 blots as each 4ml of working solution is suitable for 1 mini blot (8 x 7.5cm).
In addition, picoLUCENT™ PLUS-HRP is also supplied in a kit format, containing our non-animal protein blocking agent (NAP-BLOCKER™) and wash buffer (femto-TBST™). The picoLUCENT™ PLUS-HRP kits allow detection of low picogram levels (10-12) of antigens with low noise (signal/background) ratio. The kit reagents are sufficient for 25 mini blots or 1,500cm2 of PVDF or nitrocellulose membrane.
The chemiluminescence light emission can be recorded by a short exposure to autoradiography films