Accucore™ XL C8 Drop-in Guard Cartridges, Thermo Scientific
Supplier: Thermo Scientific
74204-014001 74204-013001 74204-012101
10038-658EA 991.23 CAD
10038-658 10038-660 10038-662
Accucore™ XL C8 Drop-in Guard Cartridges, Thermo Scientific
Chromatography Columns
New Design protects the main column with minimized impact on retention or efficiency.
  • Economical and Easy To Use
  • Compatible with Uniguard™ Direct-Connection and Stand-Alone Holders
  • Similar Selectivity to C18, with Lower Retention
  • Recommended for Analytes with Moderate Hydrophobicity
Offer easy, economical protection of the main column with minimized impact on retention or efficiency. Based on Core Enhanced Technology™ using 4 µm solid core particles, Accucore™ XL C8 Drop-in Guard Cartridges allow users of conventional HPLC methods to enjoy performance far beyond that of columns packed with 5, 4, or even 3 µm fully porous particles.