Standard Tin Capsule Kit, PerkinElmer
Supplier: PerkinElmer
N2411255 N2411362
CA11007-692PK 375.03 CAD
CA11007-692 CA97013-058
Standard Tin Capsule Kit, PerkinElmer
Tin Capsules aid in the combustion of samples.
  • Easy to use
  • Aids breakdown of samples
  • Used to analyze a wide range of sample types
As it burns, tin releases heat, causing the sample to break down more quickly and more completely. Tin capsules are routinely used to analyze polymers, soils, coal, dry dairy products, plant leaves, paper, textiles, calibration standards and many organic and pharmaceutical products. To use, simply place the sample in the tin capsule and fold the capsule closed.