VWR® Cell Scrapers
Supplier: VWR International
10062-904 CSC012025 10062-908 10062-910
10062-904CS 367.22 CAD
10062-904 10062-906 10062-908 10062-910
VWR® Cell Scrapers
Cell Scrapers
Specifically designed to make collecting of cells easier and more effective.
  • Cross-ribbed handle in ABS provides a greater rigidity and ensures a better control while scraping cells
  • Thin, flexible TPE blade prevents damage to cells
  • Individually wrapped
  • Sterilized by electron beam irradiation
  • Non pyrogenic
Available with a choice of blade positions; scraper for use in flasks, or lifter for use in harvesting cells (especially stem cells) in dishes.