Cover Glasses, Rectanglular, Corning®
Supplier: Corning
2975-223 2975-224 2975-225 2975-244 2975-245
89239-708CS 423.57 CAD
89239-708 89239-710 89239-712 89239-716 89239-718
Cover Glasses, Rectanglular, Corning®
Cover Glasses
Special, optically clear, water-white, borosilicate cover glass is resistant to surface attachment or weathering and will remain clear for extended periods of time.
  • Designed for use in general laboratory microscopy
Nonsterile.