Traceable® Certified Reference Material (CRM) pH Buffers, Control Company
Supplier: Antyila Scientific
00651-00 00651-02 00655-46 00651-04 00655-30 00655-32 00655-36 00655-34
10053-726EA 61.67 CAD
10053-726 10053-728 10053-730 10053-732 10053-734 10053-736 10053-738 10053-740
Traceable® Certified Reference Material (CRM) pH Buffers, Control Company
Buffers pH Calibration Buffers
Traceable® pH Standards, Certified Reference Materials, are 100% compatible with all instruments and probes
Certified Reference Materials meet Federal/State/local agencies’ strictest mandates and deliver exact calibration results for any pH meter. Select values are manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation. All Certified Reference Materials feature resistance to temperature changes and long-term buffer stability.
ISO 17025 (Calibration Laboratory) and ISO Guide 34 (Certified Reference Material Producer) provide highest achievable levels of product production, documentation, and accuracy. Additional accreditations include ISO 31 (content) and ISO 35 (statistical analysis). ISO 9001 ensures that world-class product quality standards are always met.
To assure accuracy an individually serial-numbered Traceable® Certificate is supplied to indicate traceability to standards provided by NIST (National Institute of Standards and Technology) and/or a National Standards Laboratory. Each 100 ml container is supplied with an individual temperature compensation chart, traceability information, and Traceable® Certificate.
Accuracy for the Traceable® Buffers at 25°C is ±0.010 pH.
Ordering information: Each 100 mL container is supplied with an individual temperature compensation chart, traceability information, and Traceable® Certificate.