Snap Caps, with Single Thumb Tab, Globe Scientific
Supplier: Globe Scientific
113146Y 113148A 113148B 113146A 113146B 113146G 113146L 113146R 113146W 113148G 113148L 113148R 113148W 113148Y
10040-008PK 73.18 CAD
10040-008 10040-010 10040-012 10040-014 10040-016 10040-018 10040-020 10040-022 10040-024 10040-026 10040-028 10040-030 10040-032 10040-034
Snap Caps, with Single Thumb Tab, Globe Scientific
Closures Test/Sample Tube Closures
These colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes, and plastic culture tubes
  • Low density polyethylene (LDPE)
  • Assorted colors available
  • Fits both glass and plastic tubes
Each cap is precision molded from low-density polyethylene, and maintains a tight seal during centrifugation, agitation, and refrigeration procedures. They are available in bright attractive colors for sample identification purposes. The caps feature a single thumb tab for easy removal of the cap. This feature also allows the tubes toMore Product Information