TraceGOLD TG-17SiLMS GC Columns, Thermo Scientific
Supplier: Thermo Scientific
26072-2750 26072-5780 26072-1540 26072-1430
10055-446EA 970.66 CAD
10055-446 10057-144 10057-150 10057-154
TraceGOLD TG-17SiLMS GC Columns, Thermo Scientific
Chromatography Columns
Optimize separation when analyzing active environmental compounds.
Ideal for separation of active environmental compounds such as PAH's. Superior inertness ensures excellent peak shape and sensitivity.