TraceGOLD TG-5SiLMS GC Columns, Thermo Scientific
Supplier: Thermo Scientific
26096-1300 26096-1420 26096-1430 26096-1540 26096-2230 26096-2240 26096-2760 26096-2960 26096-2970 26096-3080 26096-3360 26096-5780
10056-424EA 1173.14 CAD
10056-424 10056-602 10056-608 10056-610 10056-614 10056-618 10056-624 10056-630 10056-632 10056-634 10056-636 10056-638 10056-642
TraceGOLD TG-5SiLMS GC Columns, Thermo Scientific
Chromatography Columns
Incorporate phenyl groups in the polymer backbone for improved thermal stability, reduced susceptibility to oxidation.