Tygon S3™ E-LFL Long Flex Life Pump Tubing, Saint-Gobain Life Sciences
Supplier: Saint Gobain Life Sciences
AVX42003 AVX42019 AVX42029 AVX06057
89305-340EA 286.88 CAD
89305-340 89305-342 89305-348 89305-352 89305-356
Tygon S3™ E-LFL Long Flex Life Pump Tubing, Saint-Gobain Life Sciences
Tubing Transfer Tubing
Non-DEHP Tygon S3™ E-LFL Tubing for laboratory, food and beverage, and biopharmaceutical applications is now phthalate-free.
Tygon S3™ E-LFL Tubing offers the longest flex life of any clear Tygon® tubing, and reduces downtime due to pump failure. It provides broad chemical resistance and is excellent for laboratory applications. Contains no BPA or phthalates.
Saint-Gobain is proud to be among the first companies to offer sustainable flexible tubing products. The bio-based Tygon S3™ line combines high-performance standards with eco-friendly tubing design. This new non-DEHP tubing is theMore Product Information
Order Now
SPECIFICATIONS
Model 89305-340 89305-342 89305-348 89305-352 89305-356
Environmentally Preferable
Durometer hardness Shore A, 56 Shore A, 56 Shore A, 56 Shore A, 56 Shore A, 56
Max. vacuum 29.9" 29.9" 29.9" 29.9" 29.9"
Working pressure 50.0 psi 34.0 psi 35.0 psi 25.0 psi 24.0 psi