TraceGOLD TG-1MS GC Columns, Thermo Scientific
Supplier: Thermo Scientific
26099-0360 26099-2760 26099-2940 26099-5680 26099-3530 26099-3590 26099-4840
10056-748EA 1372.18 CAD
10056-748 10056-750 10056-752 10056-754 10056-756 10056-760 10056-762 10056-764 10056-766 10056-770 10056-772 10056-774 10056-776 10056-778 10056-780 10056-784 10056-786 10056-788 10056-790 10056-792 10056-794 10056-796 10056-798 10056-802 10056-804 10056-806 10056-808 10056-814 10841-572 10841-578 10841-584
TraceGOLD TG-1MS GC Columns, Thermo Scientific
Chromatography Columns
Exceptionally low bleed for optimal signal-to-noise ratio, sensitivity and mass spec integrity.
  • Non polar, 100% dimethyl polysiloxane
  • Equivalent to USP G2
  • Exceptionally low bleed
These columns are recommended for hydrocarbons, GROs, drugs of abuse and PCBs.