Nalgene® Economy Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS2185-0004 DS2185-0008 DS2185-0016 DS2185-0032 2189-0001 2189-0002 2189-0004 2189-0008 2189-0016 2189-0032
16172-140CS 353.57 CAD
16172-140 16172-144 16172-146 16172-148 16172-150 16172-160 16172-162 16172-164 16172-166 16172-168 16172-170
Nalgene® Economy Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Economical, single-use, break-resistant bottles feature thinner walls and are more flexible than original Nalgene® bottles.
  • Excellent chemical resistance to most acids, bases, and alcohols
  • Available in natural and amber finishes
  • Guaranteed leakproof
Ideal for short-term, general-purpose laboratory and field applications, such as collecting samples, or for storing working solutions like buffers, bench reagents, and diluted chemical aliquots. Amber bottles reduce UV light transmission for protection of light-sensitive liquids.
Certifications: Amber bottles comply with U.S. Pharmacopoeia 23 requirements for maximum light transmission.
Delivery information: Supplied with polypropylene screw closures.
Caution: Not recommended for hazardous and critical material containment.