FLIPR Calcium 6 Assay Kits, Molecular Devices
Supplier: MOLECULAR DEVICES LLC. CA
R6133 R6134 R8194 R8190 R8191 R8195 R8192 R8193 R8196 R6133 R6134 R8194 R8190 R8191 R8195 R8192 R8193 R8196
10048-800EA 435.67 CAD
10048-800 10048-802 10048-804 10048-806 10048-808 10048-810 10048-812 10048-814 10048-816 CA10048-800 CA10048-802 CA10048-804 CA10048-806 CA10048-808 CA10048-810 CA10048-812 CA10048-814 CA10048-816
FLIPR Calcium 6 Assay Kits, Molecular Devices
Assays Cellular Assays
Built as the most comprehensive calcium portfolio, the FLIPR® Calcium Assay Kits measure changes in intracellular calcium during drug discovery and research.
  • Maximizes fluorescent signal
  • Reduces well-to-well variability
  • Universal mix-and-read protocols
  • Superior signal-to-noise ratio
  • Pre-optimized and validated protocols
They deliver pre-optimized, homogeneous, fluorescence-based formulations to expedite assay development and screening of GPCR and ion channel targets.
FLIPR® Calcium Assay Kits maximize fluorescent signal from a wide range of cell types by reducing cell wash artifacts. They reduce well-to-well variability, improving assay quality (Z'-factor) and reliability (CV %) of high-throughput screens. Universal mix-and-read protocol accelerates assay workflow and increases throughput. Superior signal-to-noise ratio facilitates confirmation of endogenous or transiently transfected receptor activity during assay development. Pre-optimized and validated protocols for the FLIPR® Tetra and FlexStation® 3 Systems ensure you can navigate both routine and unconventional cell lines and targets.