Water Pollution Standard, Total Petroleum Hydrocarbons (TPH) in Water, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
600 642 601
CA11020-134EA 421.89 CAD
CA11020-134 CA11020-136 CA11022-878
Water Pollution Standard, Total Petroleum Hydrocarbons (TPH) in Water, ERA (Waters)
Standards Environmental Analysis Standards
One liter whole-volume bottle is ready to analyze
Use with EPA methods 418.1, 1664, 5520 and 8440.
Order Now
SPECIFICATIONS
Environmentally Preferable