Flanged Plug Caps, Tiered, 13 mm, Globe Scientific
Supplier: Globe Scientific
118240C 118240L 118240N 118240W
89206-925EA 54.07 CAD
89206-925 89497-879 89497-881 89497-883
Flanged Plug Caps, Tiered, 13 mm, Globe Scientific
Closures Test/Sample Tube Closures
These polyethylene flanged plug caps provide exceptional leak-resistance in 13 mm test tubes, centrifuge tubes, and round cuvettes.
  • Low density polyethylene (LDPE)
  • Assorted colors available
  • Fits both glass and plastic tubes
They feature two flexible flanges that press firmly against the inside wall of the tubes, thereby providing better leak-resistance than standard plug caps. They are available in a large selection of colors for sample identification purposes.
Ordering information: Packaged in bags of 1000.