Water Pollution Standard, Nitrite, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
770 888
CA11020-092EA 326.59 CAD
CA11020-092 CA11027-930
Water Pollution Standard, Nitrite, ERA (Waters)
Standards Environmental Analysis Standards
One 15 mL screw-cap vial yields up to 2 L of this water pollution, nitrite standard after dilution.