Water Pollution Standard, Simple Nutrients, 15 ml, ERA
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
584 505
CA11020-116EA 326.59 CAD
CA11020-116 CA11020-118
Water Pollution Standard, Simple Nutrients, 15 ml, ERA
Standards Environmental Analysis Standards
One 15 mL screw-cap vial yields up to 2 L of this water pollution, simple nutrients standard after dilution.