Water Pollution Standard, Solids, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
499 241
CA11022-860EA 455.86 CAD
CA11022-860 CA11027-918
Water Pollution Standard, Solids, ERA (Waters)
Standards Environmental Analysis Standards
This whole-volume standard is ready-to-use as provided
It requires no dilution before analysis. One 500 mL whole-volume bottle of this water pollution, solids standard is ready to analyze.