Water Pollution Standard, Volatile Solids, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
913 884
CA11020-150EA 333.41 CAD
CA11020-150 CA11022-908
Water Pollution Standard, Volatile Solids, ERA (Waters)
Standards Environmental Analysis Standards
One 12 mL screw-cap vial with a solid yields 1 L of this volatile solids standard after dilution
Use with EPA method 160.4 and with Standards method 2540E.