Water Pollution Standard, Minerals, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
581 506
CA11020-086EA 489.83 CAD
CA11020-086 CA11020-088
Water Pollution Standard, Minerals, ERA (Waters)
Standards Environmental Analysis Standards
This whole-volume standard is ready-to-use as provided
It requires no dilution before analysis. One 500 mL whole-volume bottle of this water pollution, minerals standard is ready to analyze.
Specific conductance: 200 - 1200 µmhos/cm (at 25 °C)