Water Supply Standard, Unregulated Volatiles, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
One 2 mL flame-sealed ampule of this water supply, regulated volatiles standard yields in excess of 200 mL after dilution
Bromobenzene, Bromochloromethane, Bromomethane, n-Butylbenzene, sec-Butylbenzene, tert-Butylbenzene, Chloroethane, Chloromethane, 2-Chlorotoluene, 4-Chlorotoluene, Dibromomethane, 1,3-Dichlorobenzene, Dichlorodifluoromethane, 1,1-Dichloroethane, 1,3-Dichloropropane, 2,2-Dichloropropane, 1,1-Dichloropropene, cis-1,3-Dichloropropene, trans-1,3 Dichloropropene, Fluorotrichloromethane, Hexachlorobutadiene, Isopropylbenzene, 4-Isopropyltoluene, Methyl tert-butyl ether (MTBE), Naphthalene, n-Propylbenzene, 1,1,1,2-Tetrachloroethane, 1,1,2,2-Tetrachloroethane, 1,2,3-Trichlorobenzene, 1,2,3-Trichloropropane, 1,2,4-Trimethylbenzene, 1,3,5-Trimethylbenzene.
Use with EPA methods 502.2 and 524.2. This standard contains at least 60% of the analytes listed below at 2 to 50 µg/L.