Water Supply Standard, Inorganics, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
591 698
CA11020-064EA 435.5 CAD
CA11020-064 CA11020-066
Water Supply Standard, Inorganics, ERA (Waters)
Standards Environmental Analysis Standards
This whole-volume standard is ready-to-use as provided
It requires no dilution before analysis. One 500 mL whole-volume bottle of this water supply, inorganics standard is ready to analyze. This standard is also certified for sodium.
Ordering information: For a sodium proficiency test, order ERA Cat. No. 555_C.