Radiochemistry Standard, Tritium, RAD, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
812 752 752QR
CA11022-894EA 1867.81 CAD
CA11022-894 CA10344-930 CA10345-648
Radiochemistry Standard, Tritium, RAD, ERA (Waters)
Standards Environmental Analysis Standards
Sample set includes two 250 mL whole volume bottles ready to analyze as received. One Sample for Tritium analysis and one for blank correction.
Analyte: Tritium: 1000 - 24000 pCi/L