Water Supply Standard, Inorganic Disinfection #2, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
5260 5262
CA11020-062EA 459.26 CAD
CA11020-062 CA11022-722
Water Supply Standard, Inorganic Disinfection #2, ERA (Waters)
Standards Environmental Analysis Standards
One 24 mL screw-cap vial yields up to 4 L of this water supply, inorganic disinfection standard (#2) after dilution.