Water Supply Standard, Inorganic Disinfection #1, ERA
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
5270 5272
CA11020-060EA 442.27 CAD
CA11020-060 CA11022-720
Water Supply Standard, Inorganic Disinfection #1, ERA
Standards Environmental Analysis Standards
One 24 mL screw-cap vial yields up to 4 L of this water supply, inorganic disinfection standard (#1) after dilution
This standard can be defined as UCMR 3 chlorate. The diluted standard will contain chlorate at 60 to 160 µg/L.